Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(cyclopentadienyl)zirconium dichloride, 98+%

Catalog No. p-3777458
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC187920050 5 g Glass bottle
AC187920250 25 g Glass bottle
AC187921000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC187920050 Supplier Thermo Scientific Chemicals Supplier No. 187920050
Only null left
Add to Cart

CAS: 1291-32-3 | C10H10Cl2Zr | 292.32 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1291-32-3
Molecular Formula C10H10Cl2Zr
Molecular Weight (g/mol) 292.32
MDL Number MFCD00003726
Synonym Zirconocene dichloride

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point 242.0°C to 245.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 98% min. (Argentometry)
Packaging Glass bottle
Quantity 5 g
Fieser 11,51; 16,393
Solubility Information Solubility in water: hydrolysis.
Formula Weight 292.32
Percent Purity 98+%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Bis(cyclopentadienyl)zirconium dichloride
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER
EINECSNumber 215-066-8

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.