Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(cyclopentadienyl)zirconium dichloride, 99+%, Thermo Scientific Chemicals

Catalog No. AA3955822
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3955822 100 g
AA3955806 5 g
AA3955814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA3955822 Supplier Thermo Scientific Chemicals Supplier No. 03955822
Add to Cart

missing translation for 'validMainframeMsg'

Zirconocene dichloride can also be used to prepare the Negishi reagent, Cp2Zr(η2-butene) which is used as a source for Cp2Zr in oxidative cyclisation reactions. 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1291-32-3
MDL Number MFCD00003726
Synonym Zirconocene dichloride; Dichlorobis(cyclopentadienyl)zirconium

Specifications

Melting Point 242°C to 245°C
Color White
Boiling Point 124°C to 125°C (15 mmHg)
Quantity 100 g
Sensitivity Air and moisture sensitive
Solubility Information Insoluble in water.
Formula Weight 292.32
Percent Purity ≥99%
Physical Form Powder
Chemical Name or Material Bis(cyclopentadienyl)zirconium dichloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 215-066-8
RTECSNumber ZH7525000
TSCA Yes
storageNote1
  • Packaged under argon
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.