Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(cyclopentadienyl)zirconium hydridochloride, 95%, Thermo Scientific Chemicals

Catalog No. AAL1516403
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1516403 1 g
AAL1516406 5 g
AAL1516414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1516403 Supplier Thermo Scientific Chemicals Supplier No. L1516403
Add to Cart

Bis(cyclopentadienyl)zirconium hydridochloride is used in the conversion of amides to aldehydes. It is also used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bis(cyclopentadienyl)zirconium hydridochloride is used in the conversion of amides to aldehydes. It is also used as a pharmaceutical intermediate.

Solubility
Reacts with water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Handle and store under inert gas. It is sensitive to moisture, light and air. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 37342-97-5
Molecular Formula C10H11ClZr-2
Molecular Weight (g/mol) 257.872
MDL Number MFCD02089401
InChI Key VPQMECJLNPUNGN-UHFFFAOYSA-M
Synonym zirconocene chloride hydride
PubChem CID 131718033
IUPAC Name cyclopenta-1,3-diene;hydride;zirconium(2+);chloride
SMILES [H-].[CH-]1C=CC=C1.[CH-]1C=CC=C1.[Cl-].[Zr+2]

Specifications

Melting Point >300°C
Quantity 1 g
UN Number UN3131
Sensitivity Air, light, and moisture sensitive
Merck Index 14,8396
Solubility Information Reacts with water.
Formula Weight 257.87
Percent Purity 95%
Chemical Name or Material Bis(cyclopentadienyl)zirconium hydridochloride
Hazard Category H228-H261-H314-H500
Hazard Statement GHS H Statement
H228-H261-H314-H318
Flammable solid.
In contact with water releases flammable gas.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P210-P223-P231+P232-P240-P241-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P335+P334-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: WATER-REACTIVE SOLID, CORROSIVE, N.O.S.
EINECSNumber 253-479-5
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.