Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Bis(Cyclopentadienyl)Zirconium(Iv) Chloride Hydride

Catalog No. 2236705G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-367-05G 5 g
22-367-025G 25 g
22-367-01G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Danger
Catalog No. 22-367-05G Supplier Sigma Aldrich Supplier No. 2236705G
Only null left
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Bis(cyclopentadienyl)zirconium(IV) chloride hydride can be used:To convert amides to aldehydes.As a reagent for the functionalization of olefins and alkynes.As a reducing agent in the preparation of (E)-vinyl organochalcogenides.To prepare a dioxaborolane derivative, which is a key intermediate for the synthesis of bridged-nicotine analogs.

Specifications

CAS 37342-97-5
Molecular Formula C10H11ClZr
Linear Formula C10H11ClZr
MDL Number MFCD02089401
Quantity 5 g
Synonym Di(cyclopentadienyl)zirconium(IV) chloride hydride; Schwartz' reagent; Zirconocene chloride hydride
Molecular Weight (g/mol) 257.87 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.