Learn More
Strem, An Ascensus Company BIS(DI-I-PROPYLAMINO)CHLORO-5G
SDP
Catalog No. 5090109879
Encompass
Bis(di-i-propylamino)chlorophosphine, min. 97%; Size: 5g; [56183-63-2]; [(C3H7)2N]2PCl; Mol Wt:266.80; Color & Form: white to off-white pwdr.
Catalog No. 50-901-09879 Supplier Strem, An Ascensus Company Supplier No. 1504485G
Bis(di-i-propylamino)chlorophosphine, min. 97%; Size: 5g; [56183-63-2]; [(C3H7)2N]2PCl; Mol Wt:266.80; Color & Form: white to off-white pwdr.