Learn More
Strem, An Ascensus Company CAS# 32005-36-0. 5g. Bis(dibenzylideneacetone)palladium(0) [Pd(dba)2]. MFCD00051942
SDP
Catalog No. 5090112292
Encompass
CAS# 32005-36-0. 5g. Bis(dibenzylideneacetone)palladium(0) [Pd(dba)2]. MFCD00051942. Molecular Weight: 575.00. Molecular Formula: (C6H5CH=CHCOCH=CHC6H5)2Pd. Color/form: purple pwdr. Strem# 46-0210. http://www.strem.com/catalog/v/46-0210/
Catalog No. 50-901-12292 Supplier Strem, An Ascensus Company Supplier No. 4602105G
CAS# 32005-36-0. 5g. Bis(dibenzylideneacetone)palladium(0) [Pd(dba)2]. MFCD00051942. Molecular Weight: 575.00. Molecular Formula: (C6H5CH=CHCOCH=CHC6H5)2Pd. Color/form: purple pwdr. Strem# 46-0210. http://www.strem.com/catalog/v/46-0210/