Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Bis(Dibenzylideneacetone)Palladium(0)

Catalog No. 2279945G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
500 mg
2 g
5 g
50 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-799-45G 5 g
22-799-4500MG 500 mg
22-799-42G 2 g
22-799-450G 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 22-799-45G Supplier Sigma Aldrich Supplier No. 2279945G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Bis(dibenzylideneacetone)palladium(0) (Pd(dba)2) is an air stable Pd0 complex. It is a reagent for the synthesis of allylic substituted cyclopentadienes. It is a homogeneous catalyst. It catalyzes the alkylation of allyl acetates by various nucleophiles under mild conditions.Bis(dibenzylideneacetone)palladium(0) (Pd(dba)2) was employed as catalyst in the following studies:Synthesis of isomeric 2-aryl-2,5-dihydrofurans, via Heck coupling of aryl bromides with alkenes using neopentyl phosphine ligands. Heck reaction of benzyl trifluoroacetate and 2,3-dihydrofuran phosphoramidite ligand.Allylation of stabilized anions.Cross coupling of allyl, alkenyl and aryl halides with organostannanes.Cross coupling of vinyl halides with alkenyl zinc species.Carbonylation of alkenyl and aryl halides.Employed with cyclic thiourea ligands in an efficient aerobic oxidation of alcohols to aldehydes and ketones.

Specifications

CAS 32005-36-0
Molecular Formula C34H28O2Pd
Linear Formula (C6H5CH=CHCOCH=CHC6H5)2Pd
MDL Number MFCD00051942
Quantity 5 g
Synonym Palladium(0) bis(dibenzylideneacetone); Pd(dba)2
Molecular Weight (g/mol) 575 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.