Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(dibenzylideneacetone)palladium(0), TCI America™
SDP

Catalog No. B13745G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B1374-5G 5 g
B13741G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B1374-5G Supplier TCI America Supplier No. B13745G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 32005-36-0
Molecular Formula C34H28O2Pd
MDL Number MFCD00051942
InChI Key UKSZBOKPHAQOMP-SVLSSHOZSA-N
Synonym bis dibenzylideneacetone palladium, bis dibenzylideneacetone palladium 0, pd dba 2, 1e,4e-1,5-diphenylpenta-1,4-dien-3-one; palladium, bis dibenzyldeneacetone palladium 0, tris dibenzylideneacetone dipalladium o, palladium 0 bis dibenzylideneacetone, pubchem14428
PubChem CID 6505921

Specifications

Melting Point 150°C
Quantity 5 g
SMILES C1=CC=C(C=C1)C=CC(=O)C=CC2=CC=CC=C2.C1=CC=C(C=C1)C=CC(=O)C=CC2=CC=CC=C2.[Pd]
IUPAC Name (1E,4E)-1,5-diphenylpenta-1,4-dien-3-one;palladium
Molecular Weight (g/mol) 575.016
Formula Weight 575.02
Physical Form Crystalline Powder
Chemical Name or Material Bis(dibenzylideneacetone)palladium(0)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.