Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(dibenzylideneacetone)palladium(0)

Catalog No. AA1276406
Change view
Click to view available options
Quantity:
0.25 g
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1276406 5 g
AA1276401 0.25 g
AA1276403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA1276406 Supplier Thermo Scientific Chemicals Supplier No. 01276406
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 32005-36-0 | C34H28O2Pd | 575.02 g/mol

Hydrogenation, isomerization, carbonylation, oxidation, C-C bond formation.Bis(dibenzylideneacetone)palladium(0) is used as a reagent for the preparation of allylic substituted cyclopentadienes. It acts as a homogeneous catalyst as well as in the alkylation of allyl acetates by various nucleophiles. Further, it is used in Suzuki reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32005-36-0
Molecular Formula C34H28O2Pd
Molecular Weight (g/mol) 575.02
MDL Number MFCD00051942
InChI Key UKSZBOKPHAQOMP-SVLSSHOZSA-N
Synonym bis dibenzylideneacetone palladium, bis dibenzylideneacetone palladium 0, pd dba 2, 1e,4e-1,5-diphenylpenta-1,4-dien-3-one; palladium, bis dibenzyldeneacetone palladium 0, tris dibenzylideneacetone dipalladium o, palladium 0 bis dibenzylideneacetone, pubchem14428
PubChem CID 6505921
IUPAC Name (1E,4E)-1,5-diphenylpenta-1,4-dien-3-one;palladium
SMILES [Pd].O=C(\C=C\C1=CC=CC=C1)/C=C/C1=CC=CC=C1.O=C(\C=C\C1=CC=CC=C1)/C=C/C1=CC=CC=C1

Specifications

Melting Point 135°C (decomposition)
Color Red to Brown
Quantity 5 g
Sensitivity Air and moisture sensitive
Solubility Information Slightly soluble in CH2Cl2,CHCl3 and benzene.Soluble in dichloromethane,chloroform,1,2-dichloroethane,acetone,acetonitrile and benzene. Insoluble in water.
Formula Weight 575
Physical Form Powder
Chemical Name or Material Bis(dibenzylideneacetone) palladium(0)
Hazard Category H317
Precautionary Statement P261-P272-P280g-P302+P352-P333+P313-P363-P501c
TSCA No
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.