Learn More
Medchemexpress LLC Bis(dibenzylideneacetone)palladium | 32005-36-0 | 575.00 | 1 G
SDP
Catalog No. 5000447677
Encompass_Preferred
Bis(dibenzylideneacetone)palladium is a biochemical reagent and organic compound used for life science research. It functions as a homogeneous catalyst and is involved in various chemical reactions.
  • Reagent for the preparation of allylic substituted cyclopentadienes
  • Acts as a homogeneous catalyst
  • Involved in the alkylation of allyl acetates by various nucleophiles
  • Used in Suzuki reactions
  • Intermediate for hydrogenation, isomerization, carbonylation, oxidation, and C-C bond formation
Catalog No. 50-004-47677 Supplier Medchemexpress LLC Supplier No. HYW0011371G
Bis(dibenzylideneacetone)palladium is a biochemical reagent and organic compound used for life science research. It functions as a homogeneous catalyst and is involved in various chemical reactions.
  • Reagent for the preparation of allylic substituted cyclopentadienes
  • Acts as a homogeneous catalyst
  • Involved in the alkylation of allyl acetates by various nucleophiles
  • Used in Suzuki reactions
  • Intermediate for hydrogenation, isomerization, carbonylation, oxidation, and C-C bond formation