Learn More
Strem, An Ascensus Company CAS# 137349-65-6. 1g. Bis(dicyclohexylphosphino)methane, min. 97%. MFCD00143763
SDP
Catalog No. 5090110555
Encompass
CAS# 137349-65-6. 1g. Bis(dicyclohexylphosphino)methane, min. 97%. MFCD00143763. Molecular Weight: 408.59. Molecular Formula: (C6H11)2PCH2P(C6H11)2. Color/form: white xtl. Strem# 15-9590. http://www.strem.com/catalog/v/15-9590/
Catalog No. 50-901-10555 Supplier Strem, An Ascensus Company Supplier No. 1595901G
Add to Cart
Add to Cart
CAS# 137349-65-6. 1g. Bis(dicyclohexylphosphino)methane, min. 97%. MFCD00143763. Molecular Weight: 408.59. Molecular Formula: (C6H11)2PCH2P(C6H11)2. Color/form: white xtl. Strem# 15-9590. http://www.strem.com/catalog/v/15-9590/