Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(dimethylamino)dimethylsilane 97.0+%, TCI America™
SDP

Catalog No. B2150250ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B2150250ML 250 mL
B215025ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B2150250ML Supplier TCI America Supplier No. B2150250ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3768-58-9
Molecular Formula C6H18N2Si
Molecular Weight (g/mol) 146.309
MDL Number MFCD00008296
InChI Key QULMGWCCKILBTO-UHFFFAOYSA-N
Synonym bis dimethylamino dimethylsilane, silanediamine, hexamethyl, silanediamine, n,n,n',n',1,1-hexamethyl, acmc-1ctl2, bis-dimethylamino dimethylsilane, n,n,n,n-hexamethylsilanediamine #, dimethylsilane, bis n,n-dimethylamino, n,n,n',n',1,1-hexamethylsilanediamine
PubChem CID 77384
IUPAC Name N-[dimethylamino(dimethyl)silyl]-N-methylmethanamine
SMILES CN(C)[Si](C)(C)N(C)C

Specifications

Color Yellow
Boiling Point 128°C
Quantity 250 mL
UN Number 2924
Formula Weight 146.31
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Bis(dimethylamino)dimethylsilane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.