Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(diphenylphosphino)acetylene, 97%

Catalog No. AA3927704
Change view
Click to view available options
Quantity:
0.5 g
2 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3927704 2 g
AA3927702 0.5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3927704 Supplier Thermo Scientific Chemicals Supplier No. 03927704
Add to Cart

CAS: 5112-95-8 | C26H20P2 | 394.39 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5112-95-8
Molecular Formula C26H20P2
Molecular Weight (g/mol) 394.39
MDL Number MFCD00003045
InChI Key FOWZHJNBFVLJGP-UHFFFAOYSA-N
Synonym bis diphenylphosphino acetylene, phosphine, ethynylenebis diphenyl, ethynylenebis diphenylphosphine, phosphine, 1,2-ethynediylbis diphenyl, 1,2-bis diphenylphosphino ethyne, 2-diphenylphosphanylethynyl diphenyl phosphane, 2-diphenylphosphanyl ethynyl diphenylphosphane, ethyne-1,2-diylbis diphenylphosphane, acmc-20almm, pubchem6917
PubChem CID 78789
IUPAC Name 2-diphenylphosphanylethynyl(diphenyl)phosphane
SMILES C1=CC=C(C=C1)P(C#CP(C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 85°C to 87°C
Quantity 2 g
Formula Weight 394.39
Percent Purity 97%
Physical Form Crystalline
Chemical Name or Material Bis(diphenylphosphino)acetylene
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 225-842-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.