Learn More
Strem, An Ascensus Company CAS# 2071-20-7. 1g. Bis(diphenylphosphino)methane, 97%. MFCD00003537
SDP
Catalog No. NC1791346
Encompass
CAS# 2071-20-7. 1g. Bis(diphenylphosphino)methane, 97%. MFCD00003537. Molecular Weight: 384.40. Molecular Formula: (C6H5)2PCH2P(C6H5)2. Color/form: white xtl. Strem# 15-0400. http://www.strem.com/catalog/v/15-0400/
Catalog No. NC1791346 Supplier Strem, An Ascensus Company Supplier No. 1504001G
CAS# 2071-20-7. 1g. Bis(diphenylphosphino)methane, 97%. MFCD00003537. Molecular Weight: 384.40. Molecular Formula: (C6H5)2PCH2P(C6H5)2. Color/form: white xtl. Strem# 15-0400. http://www.strem.com/catalog/v/15-0400/