Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(diphenylphosphino)methane, 97%

Catalog No. AAB2111209
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2111209 10 g
AAB2111204 2 g
AAB2111218 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2111209 Supplier Thermo Scientific Chemicals Supplier No. B2111209
Add to Cart

CAS: 2071-20-7 | C25H22P2 | 384.40 g/mol

It is applied as an organic synthesis catalyst. It is also used as an intermediate in flavor and fragrances.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is applied as an organic synthesis catalyst. It is also used as an intermediate in flavor and fragrances.

Solubility
Insoluble in water.

Notes
Air sensitive. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2071-20-7
Molecular Formula C25H22P2
Molecular Weight (g/mol) 384.40
MDL Number MFCD00003537
InChI Key XGCDBGRZEKYHNV-UHFFFAOYSA-N
Synonym bis diphenylphosphino methane, dppm, methylenebis diphenylphosphine, phosphine, methylenebis diphenyl, tetraphenylmethylenediphosphine, unii-i5o2hzr38v, methanediylbis diphenylphosphine, diphenylphosphanyl methyl diphenylphosphane, 1,1-bis diphenylphosphino methane
PubChem CID 74952
IUPAC Name diphenylphosphanylmethyl(diphenyl)phosphane
SMILES C(P(C1=CC=CC=C1)C1=CC=CC=C1)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 118°C to 119°C
Quantity 10 g
Beilstein 758760
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 384.4
Percent Purity 97%
Chemical Name or Material Bis(diphenylphosphino)methane
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 218-194-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.