Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Bis-Dpeg™13-Pfp Ester

Catalog No. QBD10984 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mg
1000 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
QBD10984 100 mg
QBD10984MG 1000 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. QBD10984 Supplier Sigma Aldrich Supplier No. QBD109841000MG
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

dPEG is a registered trademark of Quanta BioDesign

Specifications

CAS 1334170-03-4
Molecular Formula C42H56F10O17
Linear Formula C42H56F10O17
MDL Number MFCD20926400
Quantity 100 mg
Molecular Weight (g/mol) 1022.87 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.