Learn More
Strem, An Ascensus Company (EtCp)2Mn, min. 98% 5g
SDP
Supplier:  Strem, An Ascensus Company 2502105G
Encompass
CAS# 101923-26-6. 5g. Bis(ethylcyclopentadienyl)manganese, min. 98%. MFCD01862452. Molecular Weight: 241.23. Molecular Formula: [(C2H5)C5H4]2Mn. Color/form: dark red liq. Strem# 25-0210. http://www.strem.com/catalog/v/25-0210/
Catalog No. 50-901-10888