Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(ethylenedithio)tetrathiafulvalene-d8 [Organic Electronic Material], TCI America™
SDP

Catalog No. B1299100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B1299100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B1299100MG Supplier TCI America Supplier No. B1299100MG

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 101751-48-8
Molecular Formula C10H8S8
Molecular Weight (g/mol) 392.703
MDL Number MFCD00070586
InChI Key LZJCVNLYDXCIBG-SVYQBANQSA-N
Synonym BEDT-TTF-d8
PubChem CID 13407544
IUPAC Name 5,5,6,6-tetradeuterio-2-(5,5,6,6-tetradeuterio-[1,3]dithiolo[4,5-b][1,4]dithiin-2-ylidene)-[1,3]dithiolo[4,5-b][1,4]dithiine
SMILES C1CSC2=C(S1)SC(=C3SC4=C(S3)SCCS4)S2

Specifications

Quantity 100 mg
Formula Weight 392.70
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Bis(ethylenedithio)tetrathiafulvalene-d8 [Organic Electronic Material]
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.