Learn More
MESA LABS BOZEMAN MFG FACILITY Mesa Labs HMV-091 Biological Indicator for VH2O2, Reliable Sterilization Assessment, Stainless Steel Carrier
SDP
Catalog No. NC2465045
Encompass
Mesa Labs HMV-091 is a biological indicator designed for validating and monitoring Vaporized Hydrogen Peroxide (VH2O2) sterilization processes in biopharmaceutical, medical device, and food industries. Its unique value lies in its robust *Geobacillus stearothermophilus* spore formulation on a stainless steel carrier, enclosed in Tyvek for reliable efficacy assessments, minimizing false positives and ensuring optimal sterilization monitoring.
Catalog No. NC2465045 Supplier MESA LABS BOZEMAN MFG FACILITY Supplier No. HMV091
Mesa Labs HMV-091 is a biological indicator designed for validating and monitoring Vaporized Hydrogen Peroxide (VH2O2) sterilization processes in biopharmaceutical, medical device, and food industries. Its unique value lies in its robust *Geobacillus stearothermophilus* spore formulation on a stainless steel carrier, enclosed in Tyvek for reliable efficacy assessments, minimizing false positives and ensuring optimal sterilization monitoring.