Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(hexamethylene)triamine 95.0+%, TCI America™
SDP

Catalog No. B181425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
400 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B181425G 25 g
B1814400G 400 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B181425G Supplier TCI America Supplier No. B181425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 143-23-7
Molecular Formula C12H29N3
Molecular Weight (g/mol) 215.39
MDL Number MFCD00010448
InChI Key MRNZSTMRDWRNNR-UHFFFAOYSA-N
Synonym bis hexamethylene triamine, bis 6-aminohexyl amine, dihexylenetriamine, 1,13-diamino-7-azatridecane, 7-azatridecane-1,13-diamine, 6,6'-diaminodihexylamine, n-6-aminohexyl hexane-1,6-diamine, 1,6-hexanediamine, n-6-aminohexyl, bishexamethylenetriamine, unii-bid902uf5v
PubChem CID 8924
IUPAC Name bis(6-aminohexyl)amine
SMILES NCCCCCCNCCCCCCN

Specifications

Melting Point 35°C
Color White-Yellow
Boiling Point 164°C
Quantity 25 g
UN Number 3259
Formula Weight 215.39
Percent Purity ≥95.0% (T)
Physical Form Crystalline Lumps
Chemical Name or Material Bis(hexamethylene)triamine
EINECSNumber (2)-2376
RTECSNumber MO1186250
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.