Learn More
Medchemexpress LLC Bis-Mal-Lysine-PEG4-TFP ester | 1426164-53-5 | 98.0% | C37H45F4N5O13 | 100 MG
SDP
Catalog No. 5000354588
Encompass_Preferred
Bis-Mal-Lysine-PEG4-TFP ester is a PEG-based PROTAC linker utilized in PROTAC synthesis. PROTACs feature two distinct ligands joined by a linker: one targeting an E3 ubiquitin ligase, the other the protein of interest. This mechanism enables PROTACs to leverage the intracellular ubiquitin-proteasome system for selective degradation of target proteins.
  • PEG-based PROTAC linker
  • Used in PROTAC synthesis
  • Enables selective degradation of target proteins
Catalog No. 50-003-54588 Supplier Medchemexpress LLC Supplier No. HY141002100MG
Bis-Mal-Lysine-PEG4-TFP ester is a PEG-based PROTAC linker utilized in PROTAC synthesis. PROTACs feature two distinct ligands joined by a linker: one targeting an E3 ubiquitin ligase, the other the protein of interest. This mechanism enables PROTACs to leverage the intracellular ubiquitin-proteasome system for selective degradation of target proteins.
  • PEG-based PROTAC linker
  • Used in PROTAC synthesis
  • Enables selective degradation of target proteins