Learn More
Medchemexpress LLC Bis-mal-peg11 | 97.3% | C38H62N4O17 | 25 MG
SDP
Catalog No. 5000352899
Encompass_Preferred
Bis-Mal-PEG11 is a PEG-based PROTAC linker that is utilized in the synthesis of PROTACs. PROTACs consist of two distinct ligands connected by a linker; one ligand targets an E3 ubiquitin ligase and the other targets a specific protein. PROTACs function by leveraging the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Utilized in the synthesis of PROTACs
  • Leverages the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-52899 Supplier Medchemexpress LLC Supplier No. 25MGHY140995
Bis-Mal-PEG11 is a PEG-based PROTAC linker that is utilized in the synthesis of PROTACs. PROTACs consist of two distinct ligands connected by a linker; one ligand targets an E3 ubiquitin ligase and the other targets a specific protein. PROTACs function by leveraging the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Utilized in the synthesis of PROTACs
  • Leverages the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins