Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(methylglycol) phthalate, MilliporeSigma™ Supelco™

Catalog No. 111012683 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-2683 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-2683 Supplier MilliporeSigma Supelco Supplier No. 36934250MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Bis(methylglycol) phthalate belongs to the class of phthalate esters widely used as plasticizers in plastic films.

Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

CAS 117-82-8
Density 1.173 g/mL (at 20°C)
Flash Point 121°C - Closed Cup
Physical Form Neat
Linear Formula C14H18O6
MDL Number MFCD00042842
Boiling Point 230°C (10 mmHg)
Synonym Bis(2-methoxyethyl) phthalate; Dimethylglycol phthalate
Type Analytical Standard
Quantity 250 mg
Formula Weight 282.29
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.