Learn More
Enzo Life Sciences bis(Methylthio)gliotoxin (5mg). CAS: 74149-38-5
SDP
Supplier:  Enzo Life Sciences BMLL1220005
Encompass_Preferred
Inhibits PAF-induced platelet aggregation (IC50=8.4 µM) and collagen-induced aggregation (IC50=84.2 µM) with no effect on arachidonic acid or ADP induced aggregation. Inhibits PAF-induced bronchoconstriction in vivo in the guinea-pig (69% at 1.0 mg/kg). Alternative name: FR-49175. Purity: ≥98% (TLC). Solubility: Soluble in DMSO (25mg/ml) and ethanol (25mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-1135