Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(methylthio)(trimethylsilyl)methane 98.0+%, TCI America™
SDP

Catalog No. B20041G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B20041G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B20041G Supplier TCI America Supplier No. B20041G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 37891-79-5
Molecular Formula C6H16S2Si
Molecular Weight (g/mol) 180.399
MDL Number MFCD01631301
InChI Key QEPMPXAUMUWNNO-UHFFFAOYSA-N
Synonym Bis(methylthio)methyltrimethylsilane, Trimethylbis(methylthio)methylsilane
PubChem CID 11008541
IUPAC Name bis(methylsulfanyl)methyl-trimethylsilane
SMILES C[Si](C)(C)C(SC)SC

Specifications

Color Yellow
Quantity 1 g
Formula Weight 180.40
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Bis(methylthio)(trimethylsilyl)methane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.