Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich bis-MPA-COOH dendrimer

Catalog No. 806072100MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
80-607-2100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 80-607-2100MG Supplier Sigma Aldrich Supplier No. 806072100MG
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • End Group Functionality: Carboxyl surface group
  • Generation: 1
  • No. Surface Groups: 6
  • Calculated Mol. Wt.: 1083 g/mol
  • bis-MPA = 2,2-bis(hydroxymethyl)propionic acid
  • Nontoxic dendrimers for drug delivery, with functional groups for conjugation or additional modifications.

Specifications

Molecular Formula C423H566O282
Linear Formula C423H566O282
Quantity 100 mg
Synonym G4-TMP-COOH
Molecular Weight (g/mol) 10162.85
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.