Learn More
Description
- Dendrimers synthesized from bis-MPA are biodegradable and have low cytotoxicity, making them well suited for use in biological research applications. These materials have been used in many applications such as signal amplification in bioassays and in drug delivery applications. After boc deprotection, amine-functionalized dendrimers can be readily used in EDC or DCC coupling reactions with carbonyl-containing compounds to yield highly functionalized materials for a variety of biomedical applications.
Specifications
Specifications
| Molecular Formula | C615H998N48O282 |
| Linear Formula | C615H998N48O282 |
| Quantity | 100 mg |
| Synonym | Bis-MPA dendrimer; NHBoc functionalized dendrimer; PFD-G4-TMP-NH-Boc |
| Molecular Weight (g/mol) | 13576.66 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.