Learn More
Research Products International Corp bis-MSB, Scintillation Grade, 5 G
SDP
Catalog No. 501978094
Encompass_Preferred
High solubility secondary fluor. Concentrations range from 0.1-0.9 g/L.
  • Melting Point: 180-181°C
  • F MAX 4160A
Catalog No. 50-197-8094 Supplier Research Products International Corp Supplier No. 111076
High solubility secondary fluor. Concentrations range from 0.1-0.9 g/L.
  • Melting Point: 180-181°C
  • F MAX 4160A