Learn More
Research Products International Corp bis-MSB [1,4-bis-(o-methylstyryl)-benzene], Scintillation Grade, 25 Grams
SDP
Catalog No. 501367974
Encompass_Preferred
High solubility secondary fluor. Concentrations range from 0.1-0.9 g/L.
  • Melting Point: 180-181°C
  • F MAX 4160A
Catalog No. 50-136-7974 Supplier Research Products International Corp Supplier No. 111077
High solubility secondary fluor. Concentrations range from 0.1-0.9 g/L.
  • Melting Point: 180-181°C
  • F MAX 4160A