Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(n-propylcyclopentadienyl)magnesium, 98+%, Thermo Scientific™

Catalog No. AA39240MD
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA39240MD 250 mg
AA3924003 1 g
AA3924006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA39240MD Supplier Thermo Scientific Chemicals Supplier No. 39240MD

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 114504-74-4
MDL Number MFCD00151384
Synonym magnesium bis 2-propylcyclopenta-1,3-dien-1-ide, magnesium 2+ ion bis 2-propylcyclopenta-1,3-dien-1-ide

Specifications

Density 0.94g/mL
Packaging Ampule
Quantity 250 mg
UN Number UN3391
Sensitivity Air & Moisture Sensitive
Formula Weight 238.66
Percent Purity ≥98%
Physical Form Crystalline
Chemical Name or Material Bis(n-propylcyclopentadienyl)magnesium
Hazard Category Danger
Hazard Statement GHS H Statement
H250-H252
Catches fire spontaneously if exposed to air.
Self-heating in large quantities
may catch fire.
Precautionary Statement GHS P Statement
P210-P222-P280-P370+P378a-P405-P422a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Do not allow contact with air.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store locked up.
Store contents under inert gas.
DOTInformation Transport Hazard Class: 4.2; Packing Group: I; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, SOLID, PYROPHORIC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.