Learn More
Medchemexpress LLC Bis-NH2-PEG2 | 929-59-9 | 99.8% | 1 G
SDP
Catalog No. 5000338825
Encompass_Preferred
Bis-NH2-PEG2, also known as PROTAC Linker 19, is a PEG-based PROTAC linker designed for use in the synthesis of PROTACs. This product is strictly for research purposes and is not intended for patient use. PROTACs are innovative molecules that function by recruiting target proteins to E3 ubiquitin ligases, leading to their ubiquitination and subsequent degradation by the proteasome system.
  • Used in the synthesis of PROTACs
  • For research purposes only
  • Utilizes the ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-38825 Supplier Medchemexpress LLC Supplier No. HY1288331G
Bis-NH2-PEG2, also known as PROTAC Linker 19, is a PEG-based PROTAC linker designed for use in the synthesis of PROTACs. This product is strictly for research purposes and is not intended for patient use. PROTACs are innovative molecules that function by recruiting target proteins to E3 ubiquitin ligases, leading to their ubiquitination and subsequent degradation by the proteasome system.
  • Used in the synthesis of PROTACs
  • For research purposes only
  • Utilizes the ubiquitin-proteasome system
  • Selectively degrades target proteins