Learn More
Medchemexpress LLC Bis-NH2-PEG2 (PROTAC Linker 19) | 929-59-9 | 99.8% | 5 G
SDP
Supplier:  Medchemexpress LLC HY1288335G
Encompass_Preferred
Bis-NH2-PEG2 (PROTAC Linker 19) is a PEG-based PROTAC linker. This compound is used in the synthesis of PROTACs, which are designed to leverage the intracellular ubiquitin-proteasome system for selective degradation of target proteins. PROTACs achieve this by linking two ligands, one targeting an E3 ubiquitin ligase and the other the protein of interest. This product is for research use only.
  • PEG-based PROTAC linker
  • Utilized in the synthesis of PROTACs
  • Facilitates selective degradation of target proteins
  • Leverages the ubiquitin-proteasome system
  • For research use only
Catalog No. 50-003-38444