Learn More
Medchemexpress LLC Bis-NH2-PEG2 (protac linker 19) | 929-59-9 | 99.82% | 500 MG
SDP
Supplier:  Medchemexpress LLC HY128833500MG
Encompass_Preferred
Bis-NH2-PEG2 (PROTAC Linker 19) is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs operate by employing two distinct ligands connected by a linker, one targeting an E3 ubiquitin ligase and the other binding to the target protein. This mechanism leverages the intracellular ubiquitin-proteasome system for the selective degradation of target proteins.
  • Used in the synthesis of PROTACs
  • Utilizes a PEG-based linker
  • Exploits the ubiquitin-proteasome system
  • Enables selective degradation of target proteins
Catalog No. 50-003-40306