Learn More
Cayman Chemical bis-ANS (potassium salt), Purity: ≥95%, CAS: 65664-81-5, 1 mg
SDP
Catalog No. 502987317
Encompass_Preferred
A high-affinity non-covalent extrinsic fluorescent dye used to analyze protein conformation; ex/em = 390/523 (free) and 484-496 nm (bound), respectively; labels mechanically damaged neurons; inhibits microtubule assembly
Catalog No. 50-298-7317 Supplier Cayman Chemical Supplier No. 218201 MG
A high-affinity non-covalent extrinsic fluorescent dye used to analyze protein conformation; ex/em = 390/523 (free) and 484-496 nm (bound), respectively; labels mechanically damaged neurons; inhibits microtubule assembly