Learn More
Medchemexpress LLC Bis-PEG1-C-PEG1-CH2COOH | 2358775-67-2 | 95.0% | 10 MG
SDP
Catalog No. 5000338746
Encompass_Preferred
Bis-PEG1-C-PEG1-CH2COOH (PROTAC Linker 26) is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Peg-based protac linker
  • Used in the synthesis of protacs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-38746 Supplier Medchemexpress LLC Supplier No. HY12884710MG
Bis-PEG1-C-PEG1-CH2COOH (PROTAC Linker 26) is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Peg-based protac linker
  • Used in the synthesis of protacs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins