Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Bis-PEG1-C-PEG1-CH2COOH | 2358775-67-2 | 95.0% | 25 MG
SDP

Catalog No. 5000341636
Encompass_Preferred

Bis-PEG1-C-PEG1-CH2COOH, also known as PROTAC Linker 26, is a PEG-based PROTAC linker. It is utilized in the synthesis of PROTACs, which are compounds designed to exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins. PROTACs achieve this by connecting two different ligands, one for an E3 ubiquitin ligase and another for the target protein, via a linker.

  • PEG-based PROTAC linker
  • Useful in the synthesis of PROTACs
  • Facilitates selective degradation of target proteins
  • Purity of 95.0%
  • Molecular weight of 350.40
  • Chemical formula C16H30O8
  • Appears as an off-white to yellow oil
  • Soluble in DMSO up to 100 mg/mL
  • Recommended storage at -20°C for pure form

Catalog No. 50-003-41636 Supplier Medchemexpress LLC Supplier No. 25MGHY128847
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Bis-PEG1-C-PEG1-CH2COOH, also known as PROTAC Linker 26, is a PEG-based PROTAC linker. It is utilized in the synthesis of PROTACs, which are compounds designed to exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins. PROTACs achieve this by connecting two different ligands, one for an E3 ubiquitin ligase and another for the target protein, via a linker.

  • PEG-based PROTAC linker
  • Useful in the synthesis of PROTACs
  • Facilitates selective degradation of target proteins
  • Purity of 95.0%
  • Molecular weight of 350.40
  • Chemical formula C16H30O8
  • Appears as an off-white to yellow oil
  • Soluble in DMSO up to 100 mg/mL
  • Recommended storage at -20°C for pure form

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.