Learn More
Medchemexpress LLC Bis-PEG1-C-PEG1-CH2COOH | 2358775-67-2 | MFCD22381072 | 95.0% | 350.40 | C16H30O8 | 5 MG
SDP
Catalog No. 5000263110
Encompass_Preferred
Bis-PEG1-C-PEG1-CH2COOH is a PEG-based bifunctional linker bearing a terminal carboxylic acid; it serves as a building block for the synthesis of PROTACs and other conjugates. The material is supplied as an off-white to yellow oil, and is typically offered at research-grade purity for small-scale synthetic work.
  • PEG-based bifunctional linker for PROTAC and conjugate synthesis.
  • Terminal carboxylic acid enables standard amide or ester coupling reactions.
  • Low molecular weight building block (MW 350.4) for modular design.
  • Supplied as an off-white to yellow oil suitable for solution-phase synthesis.
  • High purity appropriate for research applications (95.0%).
  • Available in small pack sizes for screening and library synthesis.
Catalog No. 50-002-63110 Supplier Medchemexpress LLC Supplier No. HY1288475MG
Bis-PEG1-C-PEG1-CH2COOH is a PEG-based bifunctional linker bearing a terminal carboxylic acid; it serves as a building block for the synthesis of PROTACs and other conjugates. The material is supplied as an off-white to yellow oil, and is typically offered at research-grade purity for small-scale synthetic work.
  • PEG-based bifunctional linker for PROTAC and conjugate synthesis.
  • Terminal carboxylic acid enables standard amide or ester coupling reactions.
  • Low molecular weight building block (MW 350.4) for modular design.
  • Supplied as an off-white to yellow oil suitable for solution-phase synthesis.
  • High purity appropriate for research applications (95.0%).
  • Available in small pack sizes for screening and library synthesis.