Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Bis-PEG1-C-PEG1-CH2COOH (PROTAC Linker 26) | 2358775-67-2 | 95.0% | 100 MG
SDP

Catalog No. 5000340263
Encompass_Preferred

Bis-PEG1-C-PEG1-CH2COOH (PROTAC Linker 26) is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. It is intended for research use only.

  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Molecular weight: 350.40
  • CAS number: 2358775-67-2
  • Purity: 95.0%
  • Appearance: Oil
  • Color: Off-white to yellow
  • Soluble in DMSO at 100 mg/mL

Catalog No. 50-003-40263 Supplier Medchemexpress LLC Supplier No. HY128847100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Bis-PEG1-C-PEG1-CH2COOH (PROTAC Linker 26) is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. It is intended for research use only.

  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Molecular weight: 350.40
  • CAS number: 2358775-67-2
  • Purity: 95.0%
  • Appearance: Oil
  • Color: Off-white to yellow
  • Soluble in DMSO at 100 mg/mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.