Learn More
Medchemexpress LLC Bis-PEG1-C-PEG1-CH2COOH (PROTAC Linker 26) | 2358775-67-2 | 95.0% | 50 MG
SDP
Catalog No. 5000340684
Encompass_Preferred
Bis-PEG1-C-PEG1-CH2COOH, also known as PROTAC Linker 26, is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs are compounds designed to induce the degradation of target proteins by exploiting the intracellular ubiquitin-proteasome system, and they consist of two different ligands connected by a linker: one for an E3 ubiquitin ligase and the other for the target protein. This product is intended for research use only.
  • PEG-based PROTAC linker: can be used in the synthesis of PROTACs.
  • Key component for PROTACs: PROTACs contain two different ligands connected by a linker, targeting an E3 ubiquitin ligase and the protein of interest.
  • Induces protein degradation: PROTACs utilize the ubiquitin-proteasome system to selectively degrade target proteins.
Catalog No. 50-003-40684 Supplier Medchemexpress LLC Supplier No. HY12884750MG
Bis-PEG1-C-PEG1-CH2COOH, also known as PROTAC Linker 26, is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs are compounds designed to induce the degradation of target proteins by exploiting the intracellular ubiquitin-proteasome system, and they consist of two different ligands connected by a linker: one for an E3 ubiquitin ligase and the other for the target protein. This product is intended for research use only.
  • PEG-based PROTAC linker: can be used in the synthesis of PROTACs.
  • Key component for PROTACs: PROTACs contain two different ligands connected by a linker, targeting an E3 ubiquitin ligase and the protein of interest.
  • Induces protein degradation: PROTACs utilize the ubiquitin-proteasome system to selectively degrade target proteins.