Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Bis-PEG10 N-hydroxysuccinimide ester | 2221949-02-4 | 95.0% | 752.76 g/mol | C32H52N2O18 | 50 MG
SDP

Catalog No. 5000204034
Encompass_Preferred

Bis-PEG10 N-hydroxysuccinimide ester is a bifunctional PEG10-based NHS ester linker used for conjugation chemistry. It features two NHS-reactive termini separated by a ten-unit polyethylene glycol spacer, enabling efficient amine coupling, increased aqueous solubility, and flexible linker geometry for targeted conjugate assembly.

  • Two NHS ester reactive groups for amine coupling.
  • Ten-unit PEG spacer for improved solubility and flexibility.
  • Suitable for linker assembly in PROTAC and ADC synthesis.
  • Typical purity 95.0%.
  • Appearance: colorless to off-white oil.
  • Storage: pure form, -20°C; in solvent, -80°C for long-term storage.

Catalog No. 50-002-04034 Supplier Medchemexpress LLC Supplier No. HY13082450MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Bis-PEG10 N-hydroxysuccinimide ester is a bifunctional PEG10-based NHS ester linker used for conjugation chemistry. It features two NHS-reactive termini separated by a ten-unit polyethylene glycol spacer, enabling efficient amine coupling, increased aqueous solubility, and flexible linker geometry for targeted conjugate assembly.

  • Two NHS ester reactive groups for amine coupling.
  • Ten-unit PEG spacer for improved solubility and flexibility.
  • Suitable for linker assembly in PROTAC and ADC synthesis.
  • Typical purity 95.0%.
  • Appearance: colorless to off-white oil.
  • Storage: pure form, -20°C; in solvent, -80°C for long-term storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.