Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Bis-PEG10-NHS ester | 2221949-02-4 | 95.0% | 752.76 | C32H52N2O18 | 100 MG
SDP

Catalog No. 5000204289
Encompass_Preferred

Bis-PEG10-NHS ester is a PEG-based linker containing two N-hydroxysuccinimide (NHS) ester groups. It is used as a cleavable linker in the synthesis of antibody-drug conjugates (ADCs) and PROTACs to enable conjugation to primary amines.

  • Contains two NHS ester functional groups for amine conjugation.
  • Purity typically 95.0%.
  • Molecular weight approximately 752.76; formula C32H52N2O18.
  • CAS number 2221949-02-4 for unambiguous identification.
  • Available in multiple pack sizes, including 100 mg.
  • Storage: pure form at -20°C; in solvent store at -80°C for longer-term stability.

Catalog No. 50-002-04289 Supplier Medchemexpress LLC Supplier No. HY130824100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Bis-PEG10-NHS ester is a PEG-based linker containing two N-hydroxysuccinimide (NHS) ester groups. It is used as a cleavable linker in the synthesis of antibody-drug conjugates (ADCs) and PROTACs to enable conjugation to primary amines.

  • Contains two NHS ester functional groups for amine conjugation.
  • Purity typically 95.0%.
  • Molecular weight approximately 752.76; formula C32H52N2O18.
  • CAS number 2221949-02-4 for unambiguous identification.
  • Available in multiple pack sizes, including 100 mg.
  • Storage: pure form at -20°C; in solvent store at -80°C for longer-term stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.