Learn More
Medchemexpress LLC Bis-PEG10-NHS ester | 2221949-02-4 | 95.0% | 752.76 | C32H52N2O18 | 25 MG
SDP
Catalog No. 5000202724
Encompass_Preferred
Bis-PEG10-NHS ester is a PEG/alkyl/ether-based bifunctional NHS ester linker used for amine-reactive bioconjugation in the synthesis of PROTACs and antibody-drug conjugates. Supplied for research use only, it has a molecular weight of 752.76 g/mol and a listed purity of 95.0%.
  • NHS ester reactive group for amine coupling.
  • PEG10 spacer provides water solubility and flexibility.
  • Suitable for PROTAC and ADC synthesis and other bioconjugation applications.
  • Available in small milligram pack sizes for research use.
  • Storage recommendations: pure form -20°C; in solvent -80°C (6 months), -20°C (1 month).
Catalog No. 50-002-02724 Supplier Medchemexpress LLC Supplier No. HY13082425MG
Bis-PEG10-NHS ester is a PEG/alkyl/ether-based bifunctional NHS ester linker used for amine-reactive bioconjugation in the synthesis of PROTACs and antibody-drug conjugates. Supplied for research use only, it has a molecular weight of 752.76 g/mol and a listed purity of 95.0%.
  • NHS ester reactive group for amine coupling.
  • PEG10 spacer provides water solubility and flexibility.
  • Suitable for PROTAC and ADC synthesis and other bioconjugation applications.
  • Available in small milligram pack sizes for research use.
  • Storage recommendations: pure form -20°C; in solvent -80°C (6 months), -20°C (1 month).