Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Bis-PEG12-acid | 2667583-09-5 | 99.9% | C28H54O16 | 500 MG
SDP

Catalog No. 5000355378
Encompass_Preferred

Bis-PEG12-acid is a PEG-based PROTAC linker used in the synthesis of PROTACs.

  • Used as a PROTAC linker
  • Appearance is an off-white to yellow solid
  • Purity of 99.9%
  • Relevant for cancer and cancer targeted therapy research
  • For research use only

Catalog No. 50-003-55378 Supplier Medchemexpress LLC Supplier No. HY141020500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Bis-PEG12-acid is a PEG-based PROTAC linker used in the synthesis of PROTACs.

  • Used as a PROTAC linker
  • Appearance is an off-white to yellow solid
  • Purity of 99.9%
  • Relevant for cancer and cancer targeted therapy research
  • For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.