Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Bis-PEG13-NHS ester | 2221949-00-2 | MFCD21363287 | 98.0% | 884.92 | C38H64N2O21 | 10 MG
SDP

Catalog No. 5000202181
Encompass_Preferred

Bis-PEG13-NHS ester is a PEG/alkyl/ether-based bifunctional linker bearing NHS ester reactive groups for conjugation to primary amines. It is used as a cleavable linker for antibody-drug conjugates and as a linker in PROTAC synthesis to enable targeted bioconjugation and protein degradation.

  • Contains two NHS ester reactive groups for conjugation to primary amines.
  • Suitable as cleavable linker for antibody-drug conjugates and PROTAC synthesis.
  • High purity, typically 98.0%.
  • Molecular weight 884.92 and formula C38H64N2O21.
  • Store sealed, away from moisture; in solvent: -80°C (6 months), -20°C (1 month).

Catalog No. 50-002-02181 Supplier Medchemexpress LLC Supplier No. HY13082510MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Bis-PEG13-NHS ester is a PEG/alkyl/ether-based bifunctional linker bearing NHS ester reactive groups for conjugation to primary amines. It is used as a cleavable linker for antibody-drug conjugates and as a linker in PROTAC synthesis to enable targeted bioconjugation and protein degradation.

  • Contains two NHS ester reactive groups for conjugation to primary amines.
  • Suitable as cleavable linker for antibody-drug conjugates and PROTAC synthesis.
  • High purity, typically 98.0%.
  • Molecular weight 884.92 and formula C38H64N2O21.
  • Store sealed, away from moisture; in solvent: -80°C (6 months), -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.