Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Bis-PEG13-NHS ester | 2221949-00-2 | MFCD21363287 | 98.0% | 884.92 | C38H64N2O21 | 50 MG
SDP

Catalog No. 5000203247
Encompass_Preferred

Bis-PEG13-NHS ester is a bifunctional PEG-based linker containing N-hydroxysuccinimide (NHS) ester reactive groups that enable efficient amine conjugation; it is used as a cleavable linker in antibody-drug conjugate and PROTAC synthesis and for general bioconjugation chemistry.

  • contains N-hydroxysuccinimide (NHS) ester reactive groups.
  • functions as a cleavable linker for ADC and PROTAC synthesis.
  • molecular formula C38H64N2O21, molecular weight 884.92.
  • cas number 2221949-00-2 for unambiguous identification.
  • purity 98.0% suitable for research applications.
  • available in a 50 MG pack suitable for lab-scale synthesis.
  • intended for research use only; not for human or clinical use.

Catalog No. 50-002-03247 Supplier Medchemexpress LLC Supplier No. HY13082550MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Bis-PEG13-NHS ester is a bifunctional PEG-based linker containing N-hydroxysuccinimide (NHS) ester reactive groups that enable efficient amine conjugation; it is used as a cleavable linker in antibody-drug conjugate and PROTAC synthesis and for general bioconjugation chemistry.

  • contains N-hydroxysuccinimide (NHS) ester reactive groups.
  • functions as a cleavable linker for ADC and PROTAC synthesis.
  • molecular formula C38H64N2O21, molecular weight 884.92.
  • cas number 2221949-00-2 for unambiguous identification.
  • purity 98.0% suitable for research applications.
  • available in a 50 MG pack suitable for lab-scale synthesis.
  • intended for research use only; not for human or clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.