Learn More
Medchemexpress LLC Bis-PEG2-acid | 19364-66-0 | 206.19 | 10 G
SDP
Catalog No. 5000436328
Encompass_Preferred
Bis-PEG2-acid is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
Catalog No. 50-004-36328 Supplier Medchemexpress LLC Supplier No. HY11255910G
Add to Cart
Add to Cart
Bis-PEG2-acid is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker