Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Bis-PEG2-NHS ester | 65869-63-8 | 99.2% | 400.34 | C16H20N2O10 | 100 MG
SDP

Catalog No. 5000203844
Encompass_Preferred

Bis-PEG2-NHS ester is a non-cleavable two-unit polyethylene glycol (PEG) linker intended for antibody-agent conjugation (ADC) research applications. It provides NHS ester reactive groups for coupling to amines and is used in bioconjugation workflows to construct antibody-drug conjugates.

  • Non-cleavable 2-unit PEG linker for antibody-agent conjugation.
  • Purity 99.24% as reported by the manufacturer.
  • Molecular weight 400.34 g/mol.
  • Chemical formula C16H20N2O10.
  • CAS number 65869-63-8.
  • Appearance solid, white to off-white.
  • Storage: powder -20°C (stable for 3 years); in solvent -80°C (6 months) or -20°C (1 month).
  • Package size options include 100 mg, 250 mg, 1 g, 5 g, 10 g, and 50 g.
  • Manufacturer documents available: data sheet, safety data sheet, certificate of analysis, handling instructions, H NMR, RP-HPLC, and mass spectrometry data.

Catalog No. 50-002-03844 Supplier Medchemexpress LLC Supplier No. HY130088100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Bis-PEG2-NHS ester is a non-cleavable two-unit polyethylene glycol (PEG) linker intended for antibody-agent conjugation (ADC) research applications. It provides NHS ester reactive groups for coupling to amines and is used in bioconjugation workflows to construct antibody-drug conjugates.

  • Non-cleavable 2-unit PEG linker for antibody-agent conjugation.
  • Purity 99.24% as reported by the manufacturer.
  • Molecular weight 400.34 g/mol.
  • Chemical formula C16H20N2O10.
  • CAS number 65869-63-8.
  • Appearance solid, white to off-white.
  • Storage: powder -20°C (stable for 3 years); in solvent -80°C (6 months) or -20°C (1 month).
  • Package size options include 100 mg, 250 mg, 1 g, 5 g, 10 g, and 50 g.
  • Manufacturer documents available: data sheet, safety data sheet, certificate of analysis, handling instructions, H NMR, RP-HPLC, and mass spectrometry data.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.