Learn More
Medchemexpress LLC Bis-PEG21-NHS ester | 2221948-98-5 | 98.0% | 1 G
SDP
Catalog No. 5000340660
Encompass_Preferred
Bis-PEG21-NHS ester is a versatile PEG/Alkyl/ether-based linker. It serves as a PROTAC linker for the synthesis of PROTACs, which are compounds designed to recruit target proteins to E3 ubiquitin ligases for degradation. Additionally, this molecule functions as a cleavable ADC linker, essential for the construction of antibody-drug conjugates (ADCs).
  • PEG/Alkyl/ether-based linker structure
  • Enables synthesis of PROTACs
  • Functions as a cleavable ADC linker
  • For research use only
Catalog No. 50-003-40660 Supplier Medchemexpress LLC Supplier No. HY1308271G
Bis-PEG21-NHS ester is a versatile PEG/Alkyl/ether-based linker. It serves as a PROTAC linker for the synthesis of PROTACs, which are compounds designed to recruit target proteins to E3 ubiquitin ligases for degradation. Additionally, this molecule functions as a cleavable ADC linker, essential for the construction of antibody-drug conjugates (ADCs).
  • PEG/Alkyl/ether-based linker structure
  • Enables synthesis of PROTACs
  • Functions as a cleavable ADC linker
  • For research use only