Learn More
Medchemexpress LLC Bis-PEG21-NHS ester | 2221948-98-5 | 98.0% | 1237.34 | C54H96N2O29
SDP
Catalog No. 5000203498
Encompass_Preferred
Bis-PEG21-NHS ester is a polyethylene glycol (PEG)-based bifunctional N-hydroxysuccinimide (NHS) ester linker used for bioconjugation. It provides a hydrophilic PEG21 spacer with two NHS-activated ester termini for conjugation to primary amines and is used in the synthesis of antibody-drug conjugates and PROTAC linkers. Typical purity is 98.0% and molecular weight is 1,237.34.
  • Bifunctional PEG linker with two NHS ester termini.
  • Hydrophilic PEG21 spacer to improve solubility.
  • Suitable for ADC and PROTAC linker synthesis.
  • High purity (98.0%) for research applications.
  • Available in multiple pack sizes for synthetic workflows.
Catalog No. 50-002-03498 Supplier Medchemexpress LLC Supplier No. HY1308275MG
Bis-PEG21-NHS ester is a polyethylene glycol (PEG)-based bifunctional N-hydroxysuccinimide (NHS) ester linker used for bioconjugation. It provides a hydrophilic PEG21 spacer with two NHS-activated ester termini for conjugation to primary amines and is used in the synthesis of antibody-drug conjugates and PROTAC linkers. Typical purity is 98.0% and molecular weight is 1,237.34.
  • Bifunctional PEG linker with two NHS ester termini.
  • Hydrophilic PEG21 spacer to improve solubility.
  • Suitable for ADC and PROTAC linker synthesis.
  • High purity (98.0%) for research applications.
  • Available in multiple pack sizes for synthetic workflows.