Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Bis-PEG21-NHS ester | 2221948-98-5 | 98.0% | 1237.34 g/mol | C54H96N2O29 | 25 MG
SDP

Catalog No. 5000203386
Encompass_Preferred

Bis-PEG21-NHS ester is a PEG/alkyl/ether-based linker used in the synthesis of PROTACs and as a cleavable linker for antibody-drug conjugates (ADCs). It is supplied as a solid with high purity and defined storage recommendations.

  • PEG/alkyl/ether-based linker
  • Cleavable linker suitable for ADC synthesis
  • Used in the construction of PROTAC molecules
  • High purity (98.0%)
  • Molecular weight 1237.34 g/mol
  • CAS number 2221948-98-5
  • Powder storage: -20°C for long-term stability

Catalog No. 50-002-03386 Supplier Medchemexpress LLC Supplier No. HY13082725MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Bis-PEG21-NHS ester is a PEG/alkyl/ether-based linker used in the synthesis of PROTACs and as a cleavable linker for antibody-drug conjugates (ADCs). It is supplied as a solid with high purity and defined storage recommendations.

  • PEG/alkyl/ether-based linker
  • Cleavable linker suitable for ADC synthesis
  • Used in the construction of PROTAC molecules
  • High purity (98.0%)
  • Molecular weight 1237.34 g/mol
  • CAS number 2221948-98-5
  • Powder storage: -20°C for long-term stability

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.